Recombinant Human IRAK1 (from E. coli)
Catalog # 77540-028
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:interleukin-1 receptor-associated kinase 1
- UniProtKB:P51617
- Protein/peptide name:IRAK1
- Purity:>90%, by SDS-PAGE
- Molecular weight:The protein has a calculated MW of 12 kDa.
- Sequence:MGSSHHHHHHSSGLVPRGSHMAGCPQGDTAGESSWGSGPGSRPTAVEGLALGSSASSSSEPPQIIINPARQKMVQKLALYEDGALDSLQLLSSSSLPGLGLEQDRQGPEESDEFQS
- Endotoxin level:Low
- Concentration:0.8 mg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-028