Order Entry
Canada
ContactUsLinkComponent
Recombinant Human IL2 (from E. coli)
Recombinant Human IL2 (from E. coli)
Catalog # 77539-954
Supplier:  Creative Biomart
CAS Number:  
Recombinant Human IL2 (from E. coli)
Catalog # 77539-954
Supplier:  Creative Biomart
Supplier Number:  IL2-341H
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µg
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    interleukin 2
  • Protein/peptide name:
    IL2
  • Purity:
    >95% by SDS-PAGE
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77539-954

Specifications

About this item