Order Entry
Canada
ContactUsLinkComponent
Recombinant Mouse Il17a (from E. coli)
Recombinant Mouse Il17a (from E. coli)
Catalog # 77539-904
Supplier:  Creative Biomart
CAS Number:  
undefined
Recombinant Mouse Il17a (from E. coli)
Catalog # 77539-904
Supplier:  Creative Biomart
Supplier Number:  IL17A-71M
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    25 µg
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    interleukin 17A
  • UniProtKB:
    Q62386
  • Protein/peptide name:
    Il17a
  • Purity:
    >95.0% as determined by: (a) Analysis by RP-HPLC (b) Analysis by SDS-PAGE
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77539-904

Specifications

About this item