Recombinant Human IL10 (from HEK293 cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg by the LAL method
- Biological activity:Immobilized human IL-10, his tag at 5 μg/ml (100 μl/well) can bind biotinylated human IL-10 R alpha, Fc, Avitag with a linear range of 0.2 - 10 ng/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:interleukin 10
- UniProtKB:P22301
- Protein/peptide name:IL10
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:This protein carries a polyhistidine tag at the N-terminus the protein has a calculated MW of 19.5 kDa. The protein migrates as 19 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together