Recombinant Hepatitis B Virus HBcAg (from Pichia pastoris)
77539-716
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Pichia pastoris
- Species:Hepatitis B virus (HBV)
- Size:20 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Hepatitis B Core Ag
- Protein/peptide name:HBcAg
- Molecular weight:Major immunospecific band migrating between 20 and 25 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Tested applications:Suitable for use in ELISA and western blot. Each laboratory should determine an optimum working titer for use in its particular application. Other applications have not been tested but use in such assays should not necessarily be excluded.
- Cat. no.:77539-716