Recombinant Rat GDF15 (from Mammalian cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:Rat
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg of the protein as determined by the LAL method
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:growth differentiation factor 15
- UniProtKB:Q9Z0J6
- Protein/peptide name:GDF15
- Purity:>80%
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together