Order Entry
Canada
ContactUsLinkComponent
Recombinant Human FLT3 (from HEK293 cells)
Recombinant Human FLT3 (from HEK293 cells)
Supplier:  Creative Biomart
Certificates
undefined
Recombinant Human FLT3 (from HEK293 cells)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <0.2 EU/μg by the LAL method
  • Reconstitution instructions:
    It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    fms-related tyrosine kinase 3
  • UniProtKB:
    P36888
  • Protein/peptide name:
    FLT3
  • Purity:
    >95% as determined by Tris-Bis PAGE; >95% as determined by HPLC.
  • Molecular weight:
    The protein has a predicted MW of 61. 2 kDa. Due to glycosylation, the protein migrates to 75 - 100 kDa based on bis-tris PAGE result.
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

Specifications

Frequently Bought Together