Recombinant Human FGF9 (from E. coli)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:4 °C
- Endotoxin content:<1 EU/µg of rHuFGF-9 as determined by LAL method
- Biological activity:Fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ng/ml, corresponding to a specific activity of >2.0×10⁶ IU/mg.
- Reconstitution instructions:We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in 1×PBS to a concentration of 0.1 - 1.0 mg/ml. Stock solutions should be apportioned into working aliquots and stored at ≤−20 centigrade. Further dilutions should be made in appropriate buffered solutions.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Fibroblast growth factor 9 (glia-activating factor)
- UniProtKB:P31371
- Protein/peptide name:FGF9
- Purity:>95% by SDS-PAGE and HPLC analyses
- Molecular weight:23.3 kDa
- Sequence:APLGEVGNYF GVQDAVPFGN VPVLPVDSPV LLSDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together