Recombinant Chicken FAP (from HEK293 cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Chicken
- Storage conditions:4 °C
- Endotoxin content:<1 EU/μg by LAL
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:fibroblast activation protein, alpha
- UniProtKB:A0A8V0ZGU9
- Protein/peptide name:FAP
- Purity:>95% by SDS-PAGE
- Molecular weight:85 kDa
- Sequence:HHHHHHHHKVVTTVDGPRALTLDDYLNGNFQYKTYFPYWVSDSEYLHQNQEDDIILFNVDMNYLTTIMTNSTMKQVNASNYVMSSDKYFIALESNYSKLWRYSYTASYHIYDLIYGGFVTENQLPHKIQYISWSPVGHKLAYVYQNNIYLKQSPREAPIKLTSDGKENEIFNGIPDWVYEEEMLATKYALWWSPSGKYLAYVQFNDSDIPVIEYSYFGEDQYPRKIIIPYPKAGAKNPTVKVFIVDTTNIEAFGPKEVPVPAVIASSDHYFTWLTWVTDSRVGVQWLKRIQNFSVLAICDFKENSNTWDCPENQQHIEESQTGWAGGFFVSAPYFTSDGSSYYKIFSDKNGYKHIHYINGSVENAIQITSGEWEAIYIFRVTNDAIFYSSNEFEGYPGRRNIYKISIGSKPIRKLCITCNLRKERCQYYTARFSERSKYYALICYGPGIPISTLFENESDRELRILEDNQELQSALQEIILPKEEINKLEVDGITLWYKMLIPPQFDRSKKYPLLIQVYGGPCSQNVKHTFSISWITYLASKEGIIVALVDGRGTAYQGDKILHAVYRRLGVYEVEDQISAVKKFIEMGFIDEKRIAIWGWSYGGYVTSLALGSGSGVFKCGIAVAPVSSWEYYASIYTERFMGLPVESDNLEHYKNSTVMARAKNFQNVEYLLIHGTADDNVHFQNSAQIAKALVNAQVDFQAMWYTDQNHGIPGLSSKHLYTHMTHFLKQCFSLSE
- Endotoxin level:Low
- Concentration:0.38 mg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together