Recombinant Human ERBB3 (from E. coli)
Catalog # 77539-412
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg (determined by the LAL method)
- Reconstitution instructions:Reconstitute in PBS or others
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:ERBB3 v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
- UniProtKB:P21860
- Protein/peptide name:ERBB3
- Purity:>95%
- Molecular weight:32.6 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Tested applications:Positive control, Immunogen, SDS-PAGE, WB. If bio-activity of the protein is needed, please check active protein.
- Cat. no.:77539-412