Recombinant E.coli ERBB2 (from E. coli)
Catalog # 77539-398
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:E. coli
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- Protein/peptide name:ERBB2
- Purity:>92% as determined by SDS-PAGE
- Molecular weight:(Theoretical molecular weight): ~12kDa
- Sequence:LCFVHTVPWDQLFRNPHQALLHSGNRPEEDCGLEGLVCNSLCAHGHCWGPGPTQCVNCSH FLRGQECVEECRVWKGLPREYVSDKRCLPCHPECQPQNSSETCFGSE
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-398