Recombinant Mouse Eif2ak2 (from Insect cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Insect cells
- Species:Mouse
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:eukaryotic translation initiation factor 2-alpha kinase 2
- UniProtKB:Q03963
- Protein/peptide name:Eif2ak2
- Molecular weight:56.33 KD (with GST tag)
- Sequence:MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYI ADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTH PDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKGFGQVFK AKHRIDGKRYAIKRVKYNTEKAEHEVQALAELNHVNIVQYHSCWEGVDYDPEHSMSDTSRYKTRCLFIQMEFCDK GTLEQWMRNRNQSKVDKALILDLYEQIVTGVEYIHSKGLIHRDLKPGNIFLVDERHIKIGDFGLATALENDGKSR TRRTGTLQYMSPEQLFLKHYGKEVDIFALGLILAELLHTCFTESEKIKFFESLRKGDFSNDIFDNKEKSLLKKLL SEKPKDRPETSEILKTLAEWRNISEKKKRNTC
- Endotoxin level:Low
- Concentration:0.05 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together