Recombinant E.coli DnaK (from E. coli)
Catalog # 77539-306
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:E. coli
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Chaperone Hsp70
- UniProtKB:P0A6Y8
- Protein/peptide name:DnaK
- Purity:>95% by SDS – PAGE
- Endotoxin level:Low
- Concentration:1 mg/ml (determined by Bradford assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-306