Recombinant Mouse Dlk1 (from HEK293 cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:delta-like 1 homolog
- UniProtKB:Q09163
- Protein/peptide name:Dlk1
- Purity:≥90% (SDS-PAGE)
- Endotoxin level:Low
- Concentration:~1 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together