Recombinant Human DDX53 (from E. coli)
Catalog # 77539-278
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:25 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:DEAD (Asp-Glu-Ala-Asp) box polypeptide 53
- UniProtKB:Q86TM3
- Protein/peptide name:DDX53
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-278