Recombinant Human CXCL14
Catalog # 77539-242
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Species:Human
- Size:100 µg
- Storage conditions:4 °C
- Reconstitution instructions:Recommended reconstitution in sterile phosphate buffered saline which containsat least 0.1% human or bovine serum albumin to prepare a stocksolution of no less than 20 μg/ml and vortex thoroughly.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:chemokine (C-X-C motif) ligand 14
- UniProtKB:O95715
- Protein/peptide name:CXCL14
- Sequence:H2N-SKCKCSRK GPKIRYSDVK KLEMKPKYPH CEEKMVIITT KSVSRYRGQE HCLHPKLQST KRFIKWYNAW NEKRRVYEEK(LC-Biotin)g-OH
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Tested applications:Activity assay
- Cat. no.:77539-242