Order Entry
Canada
ContactUsLinkComponent
Recombinant Human CSF2RA (from HEK293 cells)
Recombinant Human CSF2RA (from HEK293 cells)
Supplier:  Creative Biomart
Certificates
undefined
Recombinant Human CSF2RA (from HEK293 cells)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <0.05 EU/μg by the LAL method
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage)
  • UniProtKB:
    P15509
  • Protein/peptide name:
    CSF2RA
  • Purity:
    >95% as determined by Tris-Bis PAGE; >95% as determined by HPLC.
  • Molecular weight:
    The protein has a predicted MW of 37.7 kDa. Due to glycosylation, the protein migrates to 68 - 75 kDa based on bis-tris PAGE result.
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

Specifications

Frequently Bought Together