Native Human COL1 (from Neo-natal fibroblast cells)
77539-102
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Native
- Source:Neo-natal fibroblast cells
- Species:Human
- Size:1 mg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Collagen Type I
- Protein/peptide name:COL1
- Purity:It is ultrapure collagen (99.9% SDS-PAGE, 97% Type I with remainder Type III collagen). SDS-PAGE shows the typical, and banding pattern. Gradual breakdown may occur over long periods of time thus creating atypical banding patterns.
- Endotoxin level:Low
- Concentration:3.0 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-102