Recombinant Cynomolgus CMA1 (from Mammalian cells)
Catalog # 77539-094
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:Cynomolgus monkey
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg of the protein as determined by the LAL method
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:chymase 1, mast cell
- UniProtKB:P56435
- Protein/peptide name:CMA1
- Purity:>80%
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-094