Recombinant Rhesus monkey CLEC9A (from HEK293 cells)
Catalog # 77539-086
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:C-type lectin domain family 9, member A
- Protein/peptide name:CLEC9A
- Purity:>95%
- Endotoxin level:Low
- Concentration:1 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Tested applications:ELISA, WB, Immunogen.
- Cat. no.:77539-086