Recombinant Human CLCA1 (from Wheat Germ)
Catalog # 77539-078
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Wheat germ
- Species:Human
- Size:10 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:chloride channel accessory 1
- UniProtKB:A8K7I4
- Protein/peptide name:CLCA1
- Molecular weight:36.74 kDa
- Sequence:ALGGVNAARRRVIPQQSGALYIPGWIENDEIQWNPPRPEINKDDVQHKQVCFSRTSSGGSFVASDVPNAPIPDLF PPGQITDLKAEIHGGSLINLTWTAP
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Tested applications:Enzyme-linked immunoabsorbent assay, western blot (recombinant protein), antibody production, protein array.
- Cat. no.:77539-078