Recombinant Human CD33 (from HEK293 cells)
Catalog # 77538-920
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Reconstitution instructions:It is recommended that 0.5 ml of sterile phosphate-buffered saline be added to the vial.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:CD33 CD33 molecule
- UniProtKB:P20138
- Protein/peptide name:CD33
- Purity:>95% as determined by SDS-PAGE and visualized by coomassie brilliant blue
- Molecular weight:Siglec-3-Fc Chimera migrates as a broad band between 60 and 75 kDa on SDS-PAGE due to post-translation modifications, in particular glycosylation. This compares with the unmodified Siglec-3 - Fc Chimera that has a predicted molecular mass of 54 kDa.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77538-920