Recombinant Human CD22 (from HEK293 cells)
Catalog # 77538-882
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg of the protein as determined by the LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized human CD22 at 0.02 μg/ml (100 µl/well) can bind anti-human CD22 Mab with a linear range of 0.2 - 50 ng/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:CD22 molecule
- UniProtKB:P20273
- Protein/peptide name:CD22
- Purity:≥ 95% as determined by SDS-PAGE. ≥ 90% as determined by SEC-HPLC.
- Molecular weight:The recombinant human CD22 consists of 679 amino acids and predictes a molecular mass of 76.6 kDa. As a result of glycosylation, it migrates as an approximately 108 kDa band in SDS-PAGE under reducing conditions.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77538-882