Recombinant Human C3 (from HEK293 cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg by the LAL method
- Biological activity:Immobilized human complement C3, his tag at 1 μg/ml (100 μl/well) can bind human anti-C3, human IgG1 human kappa with a linear range of 0.1 - 4 ng/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:complement component 3
- UniProtKB:P01024
- Protein/peptide name:C3
- Purity:>95% as determined by SDS-PAGE. >90% as determined by SEC-MALS.
- Molecular weight:This protein carries a polyhistidine tag at the C-terminus the protein has a calculated MW of 186.8 kDa. The protein migrates as 70 kDa, 100 - 120 kDa and 160 - 180 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together