Recombinant Human BIRC5 (from E. coli)
Catalog # 77538-738
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:baculoviral IAP repeat containing 5
- UniProtKB:O15392
- Protein/peptide name:BIRC5
- Purity:>90%, by SDS-PAGE
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77538-738