Recombinant Human BCL2 (from E. coli)
Catalog # 77538-730
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:20 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:B-Cell CLL/Lymphoma 2
- UniProtKB:P10415
- Protein/peptide name:BCL2
- Purity:>80%
- Molecular weight:~51 kDa
- Endotoxin level:Low
- Concentration:0.2 μg/μl
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77538-730