Order Entry
Canada
ContactUsLinkComponent
Recombinant Human ARG2 (from E. coli)
Recombinant Human ARG2 (from E. coli)
Supplier:  Creative Biomart
Certificates
undefined
Recombinant Human ARG2 (from E. coli)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    arginase, type II
  • UniProtKB:
    P78540
  • Protein/peptide name:
    ARG2
  • Purity:
    >90%, by SDS-PAGE
  • Sequence:
    MASMTGGQQMGRGHHHHHHENLYFQGGEFVHSVAVIGAPFSQGQKRKGVEHGPAAIREAGLMKRLSSLGCHLKDF GDLSFTPVPKDDLYNNLIVNPRSVGLANQELAEVVSRAVSDGYSCVTLGGDHSLAIGTISGHARHCPDLCVVWVD AHADINTPLTTSSGNLHGQPVSFLLRELQDKVPQLPGFSWIKPCISSASIVYIGLRDVDPPEHFILKNYDIQYFS MRDIDRLGIQKVMERTFDLLIGKRQRPIHLSFDIDAFDPTLAPATGTPVVGGLTYREGMYIAEEIHNTGLLSALD LVEVNPQLATSEEEAKTTANLAVDVIASSFGQTREGGHIVYDQLPTPSSPDESENQARVRI
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Tested applications:
    May be used for in vitro ARG2 protein mediated arginine depletion in tumor microenviroment regulation study in various cancer cells with 'ProFectin' reagent based intracellular delivery of this protein (Note: Mitochondrial transit peptide was removed, so this recombinant ARG2 protein will be only located at cytoplasmic one after intracellular protein delivery ). May be used as specific protein substrate for kinase and ubiquitin (Sumo pathway) related enzyme functional screening assays. May be used for protein-protein interaction mapping. As immunogen for specific antibody production.

Specifications

Frequently Bought Together