Order Entry
Canada
Orders LinkContactUsLinkComponent
XFD700 acid

XFD700 acid

10 mg
Catalog # 77461-698
Supplier:   AAT Bioquest
Supplier #:  1799
XFD700 acid
Catalog # 77461-698
Supplier:  AAT Bioquest
CAS Number:  
XFD700 acid
XFD700 acid

10 mg

CAS Number:  
Price
$768.97
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

XFD700 is the same molecule as the Alexa Fluor® 700 (Alexa Fluor® is the trademark of ThermoFisher).

  • Stable for 12 months

Quality tested in-house

  • Molecular Weight:1102.97 g/mol
Specifications

Specifications

  • Size:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.