Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
XFD568 amine
XFD568 amine
XFD568 amine
XFD568 amine
Catalog # 77461-536
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
XFD568 amine
Catalog # 77461-536
Supplier:
AAT BIOQUEST INC
Supplier Number:
1708
CAS Number:
Specifications
Size:
1 mg
Cat. no.:
77461-536
Specifications
About this item
Product Details
:
:
850.83 g/mol
Product Details