$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
8WS maleimide [TQ8WS maleimide]
8WS maleimide [TQ8WS maleimide], Tide Quencher™
8WS maleimide [TQ8WS maleimide], Tide Quencher™
Catalog # 77461-872
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
8WS maleimide [TQ8WS maleimide], Tide Quencher™
Catalog # 77461-872
Supplier:
AAT BIOQUEST INC
Supplier Number:
2134
CAS Number:
Specifications
Size:
1 mg
Cat. no.:
77461-872
Specifications
About this item
Product Details
:
:
1000.03 g/mol
Product Details