$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
7.1WS acid [TQ7.1WS acid]
7.1WS acid [TQ7.1WS acid], Tide Quencher™
7.1WS acid [TQ7.1WS acid], Tide Quencher™
Catalog # 77461-800
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
7.1WS acid [TQ7.1WS acid], Tide Quencher™
Catalog # 77461-800
Supplier:
AAT BIOQUEST INC
Supplier Number:
2114
CAS Number:
Specifications
Size:
5 mg
Cat. no.:
77461-800
Specifications
About this item
Product Details
:
:
978.04 g/mol
Product Details