Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5.1WS amine [TQ5.1WS amine]
5.1WS amine [TQ5.1WS amine], Tide Quencher™
5.1WS amine [TQ5.1WS amine], Tide Quencher™
5.1WS amine [TQ5.1WS amine], Tide Quencher™
Catalog # 77461-774
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
5.1WS amine [TQ5.1WS amine], Tide Quencher™
Catalog # 77461-774
Supplier:
AAT BIOQUEST INC
Supplier Number:
2074
CAS Number:
Specifications
Size:
1 mg
Cat. no.:
77461-774
Specifications
About this item
Product Details
:
:
1108.11 g/mol
Product Details