Orders LinkContactUsLinkComponent
XFD488 AML-NHS ester, ReadiCleave™
1 mg
  77463-030
 :  7008
XFD488 AML-NHS ester, ReadiCleave™
  77463-030
 :  
XFD488 AML-NHS ester, ReadiCleave™
XFD488 AML-NHS ester, ReadiCleave™
1 mg
 :  
$695.29
Each(1/Itm/Bx)
ReadiCleave™ AML XFD488 contains an azidomethyl linker that can be cleaved with TCEP to remove the Alexa Fluor® 488 fluorophore from the target molecule.
  • Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
 : For Research Use Only
  •  :1340.49 g/mol
  • Size:
    1 mg