Order Entry
Canada
Orders LinkContactUsLinkComponent
SSL biotin NHS ester, ReadiCleave™

SSL biotin NHS ester, ReadiCleave™

5 mg
Catalog # 77462-174
Supplier:   AAT Bioquest
Supplier #:  3030
SSL biotin NHS ester, ReadiCleave™
Catalog # 77462-174
Supplier:  AAT Bioquest
CAS Number:  122266-55-1
SSL biotin NHS ester, ReadiCleave™
SSL biotin NHS ester, ReadiCleave™

5 mg

CAS Number:  122266-55-1
Price
534,20 $
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

ReadiCleave™ SSL biotin NHS ester is a cleavable biotinylating reagent which can efficiently introduce a biotin moiety to amine-containing biomolecules.

  • Stable for 12 months

AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.

Caution: For Research Use Only

  • CAS Number:122266-55-1
  • Molecular Weight:504.64 g/mol
Specifications

Specifications

  • Size:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.