Order Entry
Canada
ContactUsLinkComponent
Propidium iodide 1 mg/ml in aqueous solution
Propidium iodide 1 mg/ml in aqueous solution
Catalog # 77461-644
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Propidium iodide 1 mg/ml in aqueous solution
Catalog # 77461-644
Supplier:  AAT BIOQUEST INC
Supplier Number:  17585
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    5 ml
  • Cat. no.:
    77461-644

Specifications

About this item

 :
 :668.4 g/mol
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921