Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nuclear Violet™ DCS1
Nuclear Violet™ DCS1 5 mM in DMSO
Nuclear Violet™ DCS1 5 mM in DMSO
Nuclear Violet™ DCS1 5 mM in DMSO
77461-624
:
AAT BIOQUEST INC
:
Nuclear Violet™ DCS1 5 mM in DMSO
77461-624
:
AAT BIOQUEST INC
:
17549
:
Size:
0.5 ml
Cat. no.:
77461-624
:
:
744.77 g/mol