Orders LinkContactUsLinkComponent
mFluor™ Violet 545 styramide
100 Slides
Catalog # 77462-296
Supplier:   AAT Bioquest
 :  45078
mFluor™ Violet 545 styramide
Catalog # 77462-296
Supplier:  AAT Bioquest
CAS Number:  
mFluor™ Violet 545 styramide
mFluor™ Violet 545 styramide
100 Slides
CAS Number:  
Price
$520.81
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
mFluor™ Violet 545 Styramide uses the bright green fluorescent dye mFluor™ Violet 545 (Ex/Em = 393/543 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
 : For Research Use Only
Specifications
Specifications
  • Size:
    100 Slides