Order Entry
Canada
Orders LinkContactUsLinkComponent
iFluor® 660 styramide

iFluor® 660 styramide

100 Slides
Catalog # 77462-268
Supplier:   AAT Bioquest
Supplier #:  44906
iFluor® 660 styramide
Catalog # 77462-268
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 660 styramide
iFluor® 660 styramide

100 Slides

CAS Number:  
Price
$534.20
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

iFluor® 660 dyes have fluorescence excitation and emission maxima of ~660 nm and ~680 nm respectively.

  • Stable for 12 months

Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.

Caution: For Research Use Only

  • Molecular Weight:1462.94 g/mol
Specifications

Specifications

  • Size:
    100 Slides

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.