Orders LinkContactUsLinkComponent
iFluor® 633 styramide
iFluor® 633 styramide
100 Slides
  77462-278
Supplier #:  45040
iFluor® 633 styramide
  77462-278
 :  
iFluor® 633 styramide
iFluor® 633 styramide
100 Slides
 :  
$534.20
Each
Minimum order quantity is 1
iFluor® 633 Styramide is optimized to a superior replacement for Opal Polaris 650 or other spectrally similar fluorescent tyramide conjugates or TSA reagents.
  • Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Caution: For Research Use Only
  •  :1430.86 g/mol
  • Size:
    100 Slides