iFluor® 405 styramide
100 Slides
Catalog # 77462-270
Supplier: AAT Bioquest
Supplier #: 44908
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
iFluor® 405 Styramide is a superior replacement for Alexa Fluor 405 tyramide or other spectrally similar fluorescent tyramide conjugates or TSA reagents.
- Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Caution: For Research Use Only
- Molecular Weight:1174.49 g/mol
Specifications
Specifications
- Size:100 Slides