Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Helixyte™ 14
Helixyte™ 14 5 mM [Equivalent to SYBR 14]
Helixyte™ 14 5 mM [Equivalent to SYBR 14]
77461-632
:
AAT BIOQUEST INC
:
Helixyte™ 14 5 mM [Equivalent to SYBR 14]
77461-632
:
AAT BIOQUEST INC
:
17563
:
Size:
0.5 ml
Cat. no.:
77461-632
:
:
474.06 g/mol