Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
FMOC-Lys(QSY-7)-OH
FMOC-Lys(QSY-7)-OH
FMOC-Lys(QSY-7)-OH
Catalog # 77462-870
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
FMOC-Lys(QSY-7)-OH
Catalog # 77462-870
Supplier:
AAT BIOQUEST INC
Supplier Number:
5261
CAS Number:
Specifications
Size:
50 mg
Cat. no.:
77462-870
Specifications
About this item
Product Details
:
:
1008.2 g/mol
Product Details