Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Cal-520® azide

100 µg
Catalog # 77461-764
Supplier:   AAT Bioquest
 :  20613
Cal-520® azide
Catalog # 77461-764
Supplier:  AAT Bioquest
CAS Number:  
Cal-520® azide
Cal-520® azide

100 µg

CAS Number:  
Price
$768.97
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cal-520® azide is an excellent building block that can be readily used to prepare a calcium-sensitive bioconjugate for monitoring calcium change spatially for a specific target.

  • Stable for 12 months

Calcium measurement is critical for numerous biological investigations.

 : For Research Use Only

  • Molecular Weight:878.71 g/mol
Specifications

Specifications

  • Size:
    100 µg

Specifications