Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Biotin-X IDA

1 mg
Catalog # 77461-414
Supplier:   AAT Bioquest
 :  12630
Biotin-X IDA
Catalog # 77461-414
Supplier:  AAT Bioquest
CAS Number:  
Biotin-X IDA
Biotin-X IDA

1 mg

CAS Number:  
Price
$173.57
Log in for availability
Each(1/Itm/Bx)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Biotin IDA is a bifunctional reagent that can be used to detect histidine-tagged proteins immobilized.

  • Stable for 12 months

Immobilized metal affinity chromatography (IMAC) is a popular method for protein purification, particularly for recombinant proteins fused to a polyhistidine-tag.

  • Molecular Weight:515.63 g/mol
Specifications

Specifications

  • Size:
    1 mg
  • Cat. No.:
    77461-414

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.