Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AzoDye-2 CPG
AzoDye-2 CPG (1000 A)
AzoDye-2 CPG (1000 A)
AzoDye-2 CPG (1000 A)
Catalog # 77461-970
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
AzoDye-2 CPG (1000 A)
Catalog # 77461-970
Supplier:
AAT BIOQUEST INC
Supplier Number:
2430
CAS Number:
Specifications
Size:
100 mg
Cat. no.:
77461-970
Specifications
About this item
Product Details
:
Product Details