Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Atto 647N maleimide
Atto 647N maleimide
Atto 647N maleimide
Atto 647N maleimide
Catalog # 77462-154
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
Atto 647N maleimide
Catalog # 77462-154
Supplier:
AAT BIOQUEST INC
Supplier Number:
2857
CAS Number:
Specifications
Size:
1 mg
Cat. no.:
77462-154
Specifications
About this item
Product Details
:
:
868.47 g/mol
MDL Number:
MFCD07370581
Product Details