Orders LinkContactUsLinkComponent
AATOM™ 488 maleimide
AATOM™ 488 maleimide
1 mg
  77462-136
Supplier #:  2816
AATOM™ 488 maleimide
  77462-136
 :  
AATOM™ 488 maleimide
AATOM™ 488 maleimide
1 mg
 :  
$383.64
Each(1/Itm/Bx)
Minimum order quantity is 1
AATOM™ 488 is a hydrophilic, rhodamine-based fluorescent dye with exceptional water solubility.
  • Stable for 12 months
Quality tested in-house
Delivery information: RT
Packaging: Minimize light exposure
Caution: For Research Use Only
  •  :825.74 g/mol
  •  :Freezer
  • Size:
    1 mg
  • Cat. No.:
    77462-136