Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2’,3’-cGAMP-iFluor 647
2’,3’-cGAMP-iFluor 647 conjugate
2’,3’-cGAMP-iFluor 647 conjugate
2’,3’-cGAMP-iFluor 647 conjugate
Catalog # 77461-740
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
2’,3’-cGAMP-iFluor 647 conjugate
Catalog # 77461-740
Supplier:
AAT BIOQUEST INC
Supplier Number:
20322
CAS Number:
Specifications
Size:
100 µg
Cat. no.:
77461-740
Specifications
About this item
Product Details
:
:
1940.24 g/mol
Product Details