Anti-IgG Goat Polyclonal Antibody
77462-846
- Antibody type:Control
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG (H, L)
- Reactivity:Human
- Size:1 mg
- Cross adsorption:No
- Storage temperature:Refrigerated (2...8 °C); Minimize light exposure
- Shipping temperature:20
- Purification:Affinity chromatography
- Cat. no.:77462-846
- Packaging:Vial
AAT Bioquest's anti-human secondary antibodies have well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
- Purified and quality-tested for consistent results
- Suitable for multiple applications
- Purified through affinity chromatography
Multiple secondary antibodies can bind to a single primary antibody.
: Research use only.
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: IgG