Orders LinkContactUsLinkComponent
Klinger Single Slit Diffraction Experiment
Single slit diffraction experiment
Catalog # 470356-816
Supplier:   Klinger
 :  KSCISSE
Klinger Single Slit Diffraction Experiment
Catalog # 470356-816
Supplier:  Klinger
Klinger Single Slit Diffraction Experiment
Klinger Single Slit Diffraction Experiment
Single slit diffraction experiment
Price
$2,212.00
Log in for availability
Each
Product Details & Documents
Prove the Heisenberg uncertainty principle by manipulating the diffraction variables of wavelength and slit width
  • Precise optical alignment: The optical alignment of the components is attained by optical bench, the setting up time is faster and experimentation is easy
  • Digital Lux Meter with transverse saddle: The digital Lux meter enables measurement of light intensity
  • The transverse saddle helps in fine movement of Lux meter perpendicular to the direction of light
Diffraction is a phenomenon of bending of waves when it encounters obstacles or narrow opening. A basic set up to observe diffraction consists a laser, a slit, screen placed at a distance. The wave fronts are partially obstructed by the slit. The intensity distribution of the diffraction pattern consists of a series of light and dark fringes with the intensity distribution is symmetric along about the central axis. The primary peak is called the central maxima. The corresponding peaks are called secondary, tertiary maxima. This is studied using the single slit experiment.
In a single slit experiment, an alternating dark and bright pattern can be seen when light is imposed on a slit with a size corresponding to the wavelength of light. The only differences between a single slit and a double-slit experiment are the diffraction patterns and the intensity graphs.
 : Kit includes (1) optical bench set 0.8 m, (1) laser source holder, (1) adjustable collimating slit holder, (1) travelling light sensor holder, (1) data processor, (1) power supply for light source.
Specifications
  • Dimensions:
    Single slit diffraction experiment